missing translation for 'onlineSavingsMsg'
Learn More
enQuireBio™ Recombinant Rat Lumican / LUM Protein Product Code.: 16012480

enQuireBio™ Recombinant Rat Lumican / LUM Protein

Product Code. 16012480
100 μg, 100µg
Click to view available options
Quantity:
1 mg
10 μg
100 μg
200 μg
50 μg
500 μg
Unit Size:
100µg
10µg
1mg
200µg
500µg
50µg
This item is not returnable. View return policy

Product Code. 16012480

Brand: enQuireBio™ QP6324ec100ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

A DNA sequence encoding the Rattus norvegicus (Rat) Lumican / LUM, was expressed in the hosts and tags indicated.

Specifications

Accession Number NP_112312.1
Gene ID (Entrez) 81682
Name Lumican / LUM Protein
Quantity 100 μg
Regulatory Status Research Use Only
Endotoxin Concentration Not determined.
Gene Symbol Lum
Protein Length Full Length
Product Type Recombinant Protein
Cross Reactivity Rat
Species E. coli
Protein Tag His
Sequence QYYDYDAPLF MYGELSPNCA PECNCPHSYP TAMYCDDLKL KSVPMVPPGI KYLYLRNNQI DHIDEKAFEN VTDLQWLILD HNLLENSKIK GKVFSKLKQL KKLHINYNNL TESVGPLPKS LQDLQLANNK ISKLGSFDGL VNLTFIYLQH NQLKEEAVSA SLKGLKSLEY LDLSFNQMSK LPAGLPTSLL TLYLDNNKIT NIPDEYFNRF TGLQYLRLSH NELADSGVPG NSFNISSLLE LDLSYNKLKS IPTVNENLEN YYLEVNKLEK FDVKSFCKIL GPLSYSKIKH LRLDGNPLTQ SSLPPDMYEC LRVANEITVN
Buffer Tris-based buffer, 50% glycerol
Purity or Quality Grade Greater than 90% as determined by SDS-PAGE.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
enQuireBio™ Recombinant Rat Lumican / LUM Protein >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.