missing translation for 'onlineSavingsMsg'
Learn More
enQuireBio™ Recombinant Sheep IL12B / P40 Protein Product Code.: 16091590

enQuireBio™ Recombinant Sheep IL12B / P40 Protein

Product Code. 16091590
100 μg, 100µg
Click to view available options
Quantity:
1 mg
10 μg
100 μg
200 μg
50 μg
500 μg
Unit Size:
100µg
10µg
1mg
200µg
500µg
50µg
This item is not returnable. View return policy

Product Code. 16091590

Brand: enQuireBio™ QP6210ye100ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

A DNA sequence encoding the Ovis aries (Sheep) IL12B / P40, was expressed in the hosts and tags indicated.

Specifications

Accession Number NP_001009438.1
Gene ID (Entrez) 443472
Name IL12B / P40 Protein
Quantity 100 μg
Regulatory Status Research Use Only
Endotoxin Concentration Not determined.
Gene Symbol IL12B
Protein Length Full Length
Product Type Recombinant Protein
Cross Reactivity Sheep
Species Yeast
Protein Tag His
Sequence IWELEKNVYV VELDWYPNAP GETVVLTCDT PEEDGITWTS DQSSEVLGSG KTLTIQVKEF GDAGQYTCHK GGEVLSRSLL LLHKKEDGIW STDILKDQKE PKAKSFLKCE AKDYSGHFTC SWLTAISTNL KFSVKSSRGS SDPRGVTCGA ASLSAEKVSM DHREYNKYTV ECQEGSACPA AEESLPIEVV MEAVHKLKYE NYTSSFFIRD IIKPDPPKNL QLRPLKNSRQ VEVSWEYPDT WSTPHSYFSL TFCVQVQGKN KREKKLFTDQ TSAKVTCHKD ANIRVQARDR YYSSFWSEWA SVSCS
Buffer Tris-based buffer, 50% glycerol
Purity or Quality Grade Greater than 90% as determined by SDS-PAGE.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
enQuireBio™ Recombinant Sheep IL12B / P40 Protein >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.