Learn More
Gilac™ Rectangular HACCP Container with Lid
Rectangular HACCP Container with Lid is a food container for large storage volumes; lid can support the weight of heavy loads.
439.00 SEK - 1140.00 SEK
Product Code | Brand | Color | Capacity (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Color | Capacity (Metric) | Price | Quantity & Availability | |||||
17978421
|
Gilac™
G119509 |
Red | 12 L |
439.00 SEK
Each |
Estimated Shipment: 28-07-2025
Log in to see stock. |
|||||
17968561
|
Gilac™
G119518 |
Yellow | 12 L |
439.00 SEK
Each |
In Stock
Log in to see stock. |
|||||
17908621
|
Gilac™
G119612 |
White | 15 L |
528.00 SEK
Each |
Estimated Shipment: 28-07-2025
Log in to see stock. |
|||||
17988811
|
Gilac™
G119611 |
Green | 15 L |
528.00 SEK
Each |
Estimated Shipment: 28-07-2025
Log in to see stock. |
|||||
17998561
|
Gilac™
G119609 |
Red | 15 L |
528.00 SEK
Each |
Estimated Shipment: 28-07-2025
Log in to see stock. |
|||||
17938621
|
Gilac™
G119618 |
Yellow | 15 L |
528.00 SEK
Each |
Estimated Shipment: 28-07-2025
Log in to see stock. |
|||||
17958751
|
Gilac™
G119809 |
Red | 35 L |
861.00 SEK
Each |
Estimated Shipment: 28-07-2025
Log in to see stock. |
|||||
17928471
|
Gilac™
G119818 |
Yellow | 35 L |
861.00 SEK
Each |
Estimated Shipment: 28-07-2025
Log in to see stock. |
|||||
17958861
|
Gilac™
G119911 |
Green | 55 L |
1140.00 SEK
Each |
Estimated Shipment: 28-07-2025
Log in to see stock. |
|||||
Description
- HACCP rectangular food container.
- For large storage volumes, lid can support the weight of heavy loads.
- Multipurpose container.
- Rewritable identification cartridge for inventory tracking.
- No gaps nor overlaps.
- Machine washable.
- Recessed handles for better handling.
- Stackable without lid.
- Compatible with all GILAC 4 and 6 wheel dollies.
Specifications
Storage Tote | |
HDPE | |
Rectangular |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
Gilac™ Rectangular HACCP Container with Lid