Learn More
RREB1 Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 27706-1-AP-150UL
Description
The RREB1 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human RREB1. This antibody recognizes human antigen. The RREB1 antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
RREB1 | |
Polyclonal | |
Unconjugated | |
150 μL | |
6239 | |
IgG |
Western Blot, Immunohistochemistry, ELISA | |
333 μg/mL | |
Rabbit | |
Primary | |
Human |
Product Suggestions
Customers who viewed this item also viewed.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.