missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Serum amyloid P component Mouse anti-Human, Clone: 1A1D10, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 66084-1-IG-150UL
This item is not returnable.
View return policy
Description
The protein encoded by this gene is a glycoprotein, belonging to the pentraxin family of proteins, which has a characteristic pentameric organization. These family members have considerable sequence homology which is thought to be the result of gene duplication. The binding of the encoded protein to proteins in the pathological amyloid cross-beta fold suggests its possible role as a chaperone. This protein is also thought to control the degradation of chromatin. It has been demonstrated that this protein binds to apoptotic cells at an early stage, which raises the possibility that it is involved in dealing with apoptotic cells in vivo.Specifications
Serum amyloid P component | |
Monoclonal | |
0.42 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P02743 | |
APCS | |
Serum amyloid P component Fusion Protein Ag14957 | |
150 ÎĽL | |
Primary | |
Human | |
Antibody | |
IgG2b |
Immunohistochemistry (Paraffin), Western Blot | |
1A1D10 | |
Unconjugated | |
APCS | |
9.5S alpha 1 glycoprotein, amyloid P component, serum, APCS, PTX2, SAP, Serum amyloid P component | |
Mouse | |
Protein A | |
RUO | |
325 | |
-20°C | |
Liquid |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-5 of 12
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Serum amyloid P component Mouse anti-Human, Clone: 1A1D10, Proteintech > 150 ÎĽL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction