Learn More
Trajan™ Replacement Needles for 5μL eVol™ and eVol™ NMR Syringes
Cone tipped replacement needle for use with 5uL NMR Syringes.
569.00 SEK - 913.00 SEK
Specifications
Material | Stainless Steel |
---|---|
Product Type | Replacement Needle |
For Use With (Equipment) | eVol NMR Syringe (5ÎĽL) |
Product Code | Brand | Needle Gauge | Needle Point Style | Length (Metric) Needle | Diameter (Metric) Outer | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Needle Gauge | Needle Point Style | Length (Metric) Needle | Diameter (Metric) Outer | Price | Quantity & Availability | ||
15218593
|
Trajan™
036922 |
25 G | Cone | 115 mm | 0.5 mm |
569.00 SEK
Pack of 2 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
12917606
|
Trajan™
036912 |
22 G | LC | 51 mm | 0.72 mm |
615.00 SEK
Pack of 5 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
12927606
|
Trajan™
036914 |
23 G | Cone | 50 mm | 0.72 mm |
615.00 SEK
Pack of 5 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
12937606
|
Trajan™
036916 |
25 G | Bevel | 70 mm | 0.5 mm |
913.00 SEK
Pack of 5 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
12947606
|
Trajan™
036918 |
26 G | Cone | 70 mm | 0.47 mm |
913.00 SEK
Pack of 5 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
Description
- Needle length is the exposed length of needle
- Choose a different replacement needle by selecting syringe volume, needle length, and outer diameter
Specifications
Stainless Steel | |
Replacement Needle | |
eVol NMR Syringe (5ÎĽL) |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
Trajan™ Replacement Needles for 5μL eVol™ and eVol™ NMR Syringes