missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SMN-Exon7 Mouse anti-Human, Mouse, Clone: 3A8G11, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 60255-1-IG-20UL
This item is not returnable.
View return policy
Description
This antibody, 60255-1-Ig, raised against the C-terminal region (275-294 aa) encoded by the exon 7.
The SMN1 gene is part of a 500 kb inverted duplication on chromosome 5q13. This duplicated region contains at least four genes and repetitive elements which make it prone to rearrangements and deletions. The repetitiveness and complexity of the sequence have also caused difficulty in determining the organization of this genomic region. The telomeric and centromeric copies of this gene are nearly identical and encode the same protein - survival motor neuron protein. The SMN complex plays a catalyst role in the assemble of small nuclear ribonucleoproteins, the building blocks of the spliceosome. Mutations in the SMN1 gene are known to cause spinal muscular atrophy 1/2.Specifications
SMN-Exon7 | |
Monoclonal | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P97801, Q16637 | |
SMN1 | |
SMN-Exon7 Fusion Protein Ag16615 | |
20 μL | |
Primary | |
Mouse, Human | |
Antibody | |
IgG1 |
Western Blot, Immunofluorescence, Immunocytochemistry | |
3A8G11 | |
Unconjugated | |
SMN1 | |
Component of gems 1, Gemin 1, SMN, SMN1, SMN2, SMNC, SMN-Exon7, SMNT, Survival motor neuron protein, T BCD541 | |
Mouse | |
Protein G | |
RUO | |
20595, 6606 | |
-20°C | |
Liquid |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-4 of
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
SMN-Exon7 Mouse anti-Human, Mouse, Clone: 3A8G11, Proteintech > 20 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction