Learn More
Bochem™ Steel Dressing Jars with Lid and Integral Handle
359.00 SEK - 990.00 SEK
Specifications
Includes | Lid |
---|---|
Material | Stainless Steel |
Quantity | 1Each |
Shape | Round |
Product Type | Metal Dressing Jar |
Product Code | Brand | Dimensions (L x W x H) | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Dimensions (L x W x H) | Price | Quantity & Availability | ||||||
10257602
|
Bochem™
8620 |
102 dia. x 80mmH |
359.00 SEK
Each |
Estimated Shipment: 09-07-2025
Log in to see stock. |
||||||
10378662
|
Bochem™
8621 |
102 dia. x 130mmH | N/A | |||||||
10665223
|
Bochem™
8623 |
124 dia. x 165mmH |
595.00 SEK
Each |
Estimated Shipment: 04-07-2025
Log in to see stock. |
||||||
10033762
|
Bochem™
8622 |
124 dia. x 130mmH |
559.00 SEK
Each |
Estimated Shipment: 18-07-2025
Log in to see stock. |
||||||
10195262
|
Bochem™
8624 |
180 dia. x 180mmH |
990.00 SEK
Each |
Estimated Shipment: 09-07-2025
Log in to see stock. |
||||||
Specifications
Lid | |
1Each | |
Metal Dressing Jar | |
Lid |
Stainless Steel | |
Round | |
Stainless Steel |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
Bochem™ Steel Dressing Jars with Lid and Integral Handle