missing translation for 'onlineSavingsMsg'
Learn More

Buerkle™ SteriPlast™/LaboPlast™ Sampling Spoon
BIOPHARMAGREENER_CHOICE

Can be used to remove powders, granulates, pastes or liquids. The long handle makes it easy to reach the bottoms of larger containers as well and allows for sampling of drums, jerrycans and bottles with small opening diameters.

Brand:  Buerkle™ 5378-5006

 View more versions of this product

Product Code. 17290594

  • 340.00 SEK / Pack of 10
Only null left
In Stock
Add to basket
This item is not returnable. View return policy

Description

Description

  • PS, white
  • Individually packaged and optionally sterilised
  • Handle length 300 mm
  • For single use
  • Clean room manufactured (clean room class 7)
  • Complies with EU food requirements and FDA regulations
Specifications

Specifications

Sampling Spoon
Not Autoclavable
20 mL
Sterile
10
Polystyrene
Long-handled
Single-use
Complies with EU food rrequirements and FDA regulations
300 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number252214 Certificate TypeCertificate of Compliance Date18/06/2025 Catalog Number5378-5006
Lot Number244205 Certificate TypeCertificate of Compliance Date20/11/2024 Catalog Number5378-5006
Lot Number242807 Certificate TypeCertificate of Compliance Date26/07/2024 Catalog Number5378-5006
Lot Number241614 Certificate TypeCertificate of Compliance Date13/05/2024 Catalog Number5378-5006
Lot Number250278 Certificate TypeCertificate of Analysis Date12/02/2025 Catalog Number5378-5006
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Buerkle™ SteriPlast™/LaboPlast™ Sampling Spoon > Sterile

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.