Learn More
Buerkle™ SteriPlast™/LaboPlast™ Sampling Spoon
Can be used to remove powders, granulates, pastes or liquids. The long handle makes it easy to reach the bottoms of larger containers as well and allows for sampling of drums, jerrycans and bottles with small opening diameters.
253.00 SEK - 340.00 SEK
Specifications
Product Type | Sampling Spoon |
---|---|
Material | Polystyrene |
Autoclavable | Not Autoclavable |
Handle Type | Long-handled |
Disposable | Single-use |
Product Code | Brand | Capacity (Metric) | Sterility | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Sterility | Price | Quantity & Availability | |||||
17250594
![]() ![]() |
Buerkle™
5378-5002 |
5 mL | Sterile |
323.00 SEK
Pack of 10 |
In Stock
Log in to see stock. |
|||||
17240594
![]() ![]() |
Buerkle™
5378-5001 |
5 mL | Non-sterile |
253.00 SEK
Pack of 10 |
In Stock
Log in to see stock. |
|||||
17290594
![]() ![]() |
Buerkle™
5378-5006 |
20 mL | Sterile |
340.00 SEK
Pack of 10 |
In Stock
Log in to see stock. |
|||||
17280594
![]() ![]() |
Buerkle™
5378-5005 |
20 mL | Non-sterile |
268.00 SEK
Pack of 10 |
In Stock
Log in to see stock. |
|||||
Description
- PS, white
- Individually packaged and optionally sterilised
- Handle length 300 mm
- For single use
- Clean room manufactured (clean room class 7)
- Complies with EU food requirements and FDA regulations
Specifications
Sampling Spoon | |
Not Autoclavable | |
Single-use | |
10 |
Polystyrene | |
Long-handled | |
Complies with EU food rrequirements and FDA regulations | |
300 mm |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
Buerkle™ SteriPlast™/LaboPlast™ Sampling Spoon