Learn More
Sulphate Solution for Ion Chromatography, 1g/L, H2O Matrix, Fisher Chemical™
All Our Certified Reference Materials and Reagents are produced under an ISO 9001 Quality Management System, also Accredited according to ISO/IEC 17025 and Accredited according to ISO Guide 34.
686.40 SEK - 1335.00 SEK
Specifications
Solution Contains | H2O |
---|---|
Concentration | 1 g/L |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
12922205
![]() |
Fisher Chemical
A 045 100 |
100 mL |
686.40 SEK
100mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
12932205
![]() |
Fisher Chemical
A 045 250 |
250 mL |
914.00 SEK
250mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
12942205
![]() |
Fisher Chemical
A 045 500 |
500 mL |
1335.00 SEK
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Specifications
H2O | |
1 g/L |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
Sulphate Solution for Ion Chromatography, 1g/L, H2O Matrix, Fisher Chemical™