Learn More
Thirty one elements solution, 10mg/L HNO3 matrix, Fisher chemical™ Product Code.: 12906193
Thirty one elements solution, 10mg/L HNO3 matrix, Fisher chemical™
Product Code. 12906193 Shop All Fisher Chemical Products
50 mL, 50mL
Click to view available options
Quantity:
50 mL
100 mL
250 mL
Unit Size:
100mL
250mL
50mL
Product Code. 12906193
Brand: Fisher Chemical MS007050
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
All Our Certified Reference Materials and Reagents are produced under an ISO 9001 Quality Management System, also Accredited according to ISO/IEC 17025 and Accredited according to ISO Guide 34.
Thirty one elements solution - Ag, Al, As, Ba, B, Cd, Ce, Co, Cr, Cu, Dy, Er, Gd, Ho, La, Li, Lu, Mn, Nd, Ni, P, Pb, Rb Se Sm Sr, Tl, Tm, U, V, Zn for ICP/MS 10mg/L HNO3 matrix
Specifications
Chemical Name or Material Thirty-One Elements
Packaging Bottle
Analyte or Component Names Ag,Al,As,Ba,B,Cd,Ce,Co,Cr,Cu,Dy,Er,Gd,Ho,La,Li,Lu,Mn,Nd,Ni,P, Pb,Rb,Se,Sm,Sr,Tl,Tm,U,V,Zn
Physical Form Liquid
Concentration 10 mg/L
Type ICP/MS Calibration Multi-element Standard Solution
Element Ag, Al, As, Ba, B, Cd, Ce, Co, Cr, Cu, Dy, Er, Gd, Ho, La, Li, Lu, Mn, Nd, Ni, P, Pb, Rb, Se, Sm, Sr, TI, Tm, U, V, Zn
Solvent or Matrix 2% HNO3
Quantity 50 mL
Product Identifier
  • Multi-element Standard Solution for ICP. 31 Components in HNO3 2% (MS 007)
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P501-Dispose of contents/container to .
Product Content Correction
Product Title
Thirty one elements solution, 10mg/L HNO3 matrix, Fisher chemical™ >