Learn More
TNNT3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 19729-1-AP-150UL
This item is not returnable.
View return policy
Description
Troponin T is the tropomyosin-binding subunit of troponin, the thin filament regulatory complex which confers calcium-sensitivity to striated muscle actomyosin ATPase activity.Specifications
TNNT3 | |
Polyclonal | |
Unconjugated | |
TNNT3 | |
Fast skeletal muscle troponin T, TNNT3, Troponin T3 skeletal fast, Troponin T-FS | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
21957, 24838, 7140 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.23 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P09739, P45378, Q9QZ47 | |
TNNT3 | |
Peptide | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-4 of
Product Content Correction
Product Title
TNNT3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 150 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction