Learn More
Fisherbrand™ TraceableOne™ Single-Use USB Temperature Data Logger
The TraceableOne™ Single-Use USB Temperature Data Logger automatically generates PDF data report via USB port.
11085.00 SEK - 11865.00 SEK
Products 8
Product Code Brand Data Memory Temperature Range (Metric) Price Quantity & Availability  
Product Code Brand Data Memory Temperature Range (Metric) Price Quantity & Availability  
17337619
View Documents
Fisherbrand™
18004-01
16,000 Readings, 10 Day, 1 Minute Interval 2°C to 8°C
11865.00 SEK
Pack of 40
Only null left
Estimated Shipment: 05-08-2025
Add to basket
17347619
View Documents
Fisherbrand™
18004-02
16,000 Readings, 10 Day, 1 Minute Interval -20°C to -15°C
11085.00 SEK
Pack of 40
Only null left
Estimated Shipment: 08-09-2025
Add to basket
17357619
View Documents
Fisherbrand™
18004-04
16,000 Readings, 30 Day, 3 Minute Interval 2°C to 8°C This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
17367619
missing translation for 'viewDocuments'
Fisherbrand™
18004-05
16,000 Readings, 30 Day, 3 Minute Interval -20°C to -15°C This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
17377619
missing translation for 'viewDocuments'
Fisherbrand™
18004-07
16,000 Readings, 60 Day, 6 Minute Interval 2°C to 8°C
11085.00 SEK
Pack of 40
Solo quedan null
Estimated Shipment: 12-09-2025
Añadir a la cesta
17387619
missing translation for 'viewDocuments'
Fisherbrand™
18004-08
16,000 Readings, 60 Day, 6 Minute Interval -20°C to -15°C
11085.00 SEK
Pack of 40
Solo quedan null
Estimated Shipment: 02-10-2025
Añadir a la cesta
17397619
missing translation for 'viewDocuments'
Fisherbrand™
18004-10
16,000 Readings, 90 Day, 10 Minute Interval 2°C to 8°C
11085.00 SEK
Pack of 40
Solo quedan null
Estimated Shipment: 14-11-2025
Añadir a la cesta
17307629
missing translation for 'viewDocuments'
Fisherbrand™
18004-11
16,000 Readings, 90 Day, 10 Minute Interval -20°C to -15°C
11085.00 SEK
Pack of 40
Solo quedan null
Estimated Shipment: 02-10-2025
Añadir a la cesta
DescripciĂłn
DescripciĂłn
  • LED alarm indicator for visual status indication
  • Time delay of 30 minutes before logging begins for enhanced logging accuracy
  • Protective plastic pouch maintains waterproof IP67 rating
  • Bar code for in-house scanning
  • Includes NIST-Traceable certificate from our A2LA accredited (1750.01) calibration laboratory
Videos
SDS
Documentos
Documentos
Certificados
CorrecciĂłn del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
TĂ­tulo del producto
Fisherbrand™ TraceableOne™ Single-Use USB Temperature Data Logger
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
Gracias por ayudarnos a mejorar nuestra web. Su comentario ha sido enviado