Learn More
Cole-Parmer™ VapLock™ Solvent Waste Kits for Jars and Pails
VapLock™ Solvent Waste Kits for Jars and Pails collect effluent from multiple sources while minimizing harmful solvent exhaust
Brand: Cole-Parmer™ WAK100P-10
This item is not returnable.
View return policy
Description
Various cap sizes available to fit your solvent containerServices
None
Specifications
Solvent Safety | |
Polypropylene | |
1/2 in. NPT(F), 1/4 in. NPT(F), 1/4 in.-28 UNF(F) |
Solvent-Compatible Containers | |
100-415 | |
Solvent Waste Kit |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-5 of 11
Product Content Correction
Product Title
Cole-Parmer™ VapLock™ Solvent Waste Kits for Jars and Pails > PP, 100-415, 1x1/2 in NPTF, 3x1/4 in NPTF, 6x1/4 in-28 UNFF; 1/ea
Spot an opportunity for improvement?Share a Content Correction