accessibility menu, dialog, popup

UserName

Great Deals on Sample Preparation, Filtration and Storage 

Save Up to 80%

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 11089-65-9

CH3CH3ONH(R)(R)HO(R)HO(R)HO(R)O(R)(R)OH(S)OHNOHNOO(R)O(R)O(R)OH(S)OH(R)OH(R)HNCH3O(E)

CAS RN 11089-65-9

IUPAC Name: (2E)-N-[(2R,3R,4R,5R,6R)-6-{2-[(2R,3S,4R,5R)-5-(2,4-dioxo-1,2,3,4-tetrahydropyrimidin-1-yl)-3,4-dihydroxyoxolan-2-yl]-2-hydroxyethyl}-2-{[(2R,3R,4R,5S,6R)-3-acetamido-4,5-dihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy}-4,5-dihydroxyoxan-3-yl]-5-methylhex-2-enamide
Synonyms: (2E)-N-[(2R,3R,4R,5R,6R)-6-{2-[(2R,3S,4R,5R)-5-(2,4-dioxo-3H-pyrimidin-1-yl)-3,4-dihydroxyoxolan-2-yl]-2-hydroxyethyl}-2-{[(2R,3R,4R,5S,6R)-3-acetamido-4,5-dihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy}-4,5-dihydroxyoxan-3-yl]-5-methylhex-2-enamide
Molecular Weight (g/mol): 718.71
Molecular Formula: C30H46N4O16
InChi Key: ZHSGGJXRNHWHRS-PEALBESXSA-N
SMILES: CC(C)C\C=C\C(=O)N[C@@H]1[C@@H](O)[C@@H](O)[C@@H](CC(O)[C@H]2O[C@H]([C@H](O)[C@@H]2O)N2C=CC(=O)NC2=O)O[C@@H]1O[C@H]1O[C@H](CO)[C@@H](O)[C@H](O)[C@H]1NC(C)=O
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
Color 
  • (2)
  • (2)
  • (1)
Physical Form 
  • (2)
  • (2)
Percent Purity 
  • (1)
  • (2)
Type 
  • (3)

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (3)
  • (2)
  • (1)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products