accessibility menu, dialog, popup

UserName

Great Deals on Sample Preparation, Filtration and Storage 

Save Up to 80%

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 11096-82-5

ClClClClClCl

CAS RN 11096-82-5

IUPAC Name: 2,2',3,3',4,4'-hexachloro-1,1'-biphenyl
Synonyms: 2,2',3,3',4,4'-hexachloro-1,1'-biphenyl
Molecular Weight (g/mol): 360.86
Molecular Formula: C12H4Cl6
InChi Key: BTAGRXWGMYTPBY-UHFFFAOYSA-N
SMILES: ClC1=CC=C(C(Cl)=C1Cl)C1=CC=C(Cl)C(Cl)=C1Cl
Solvent or Matrix 
  • (1)
  • (1)
  • (3)
  • (3)
  • (1)
  • (1)
Concentration 
  • (2)
  • (3)
  • (3)
  • (1)
Technique 
  • (1)

Narrow Results

Narrow Results

  • (9)
  • (1)
  • (4)
  • (1)
  • (1)
  • (3)
  • (3)
  • (1)
  • (1)
  • (2)
  • (3)
  • (3)
  • (1)
  • (1)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products