accessibility menu, dialog, popup

Test

CAS RN 11141-16-5

CAS RN 11141-16-5

IUPAC Name:
Synonyms:
Molecular Weight (g/mol):
Molecular Formula:
InChi Key:
SMILES:
Quantity 
  • (8)
  • (2)
Solvent or Matrix 
  • (1)
  • (1)
  • (3)
  • (1)
  • (1)
  • (2)
Concentration 
  • (2)
  • (4)
  • (2)
For Use With (Equipment) 
  • (1)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products

Narrow Results

Narrow Results

  • (8)
  • (2)
  • (8)
  • (2)
  • (1)
  • (1)
  • (3)
  • (1)
  • (1)
  • (2)
  • (2)
  • (4)
  • (2)
  • (1)