accessibility menu, dialog, popup

UserName

Great Deals on Sample Preparation, Filtration and Storage 

Save Up to 80%

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 113975-22-7

FNI

CAS RN 113975-22-7

IUPAC Name: 2-fluoro-3-iodopyridine
Synonyms: 2-fluoro-3-iodopyridine
Molecular Weight (g/mol): 222.99
Molecular Formula: C5H3FIN
InChi Key: WCDCAXVNBOLWNO-UHFFFAOYSA-N
SMILES: FC1=NC=CC=C1I
Molecular Weight (g/mol) 
  • (1)
  • (3)
Percent Purity 
  • (1)
  • (3)
Quantity 
  • (3)
  • (1)

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (1)
  • (3)
  • (1)
  • (3)
  • (3)
  • (1)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products