CAS RN 41340-25-4
CAS RN 41340-25-4
IUPAC Namn:
2-{1,8-diethyl-1H,3H,4H,9H-pyrano[3,4-b]indol-1-yl}acetic acid
Synonymer:
etodolac etodolac, (-)-isomer ultradol etodolac (product) ramodar monosodium salt etodolac etodolac, (-)- etodolac (substance) etodolac, (+-)- etodolac, (S)-
Molekylvikt (g/mol):
287.36
Molekylär sammansättning:
C17H21NO3
InChi Key:
NNYBQONXHNTVIJ-UHFFFAOYNA-N
SMILES:
CCC1=C2NC3=C(CCOC3(CC)CC(O)=O)C2=CC=C1
1
–
4
av
4
Resultat
1
Thermo Scientific Chemicals Etodolac
CAS: 41340-25-4 Molekylformel: C17H21NO3
2
Etodolac, Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
3
Etodolac, LoGiCal
LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.
4
Etodolac, MedChemExpress
MedChemExpress Etodolac (AY-24236) is a non-steroidal anti-inflammatory compound that is a non-selective inhibitor of COX (IC50=53.5 nM)