CAS RN 2922-83-0
IUPAC Namn:
(2S)-2-amino-4-(2-aminophenyl)-4-oxobutanoic acid
Synonymer:
L-kynurenine
Molekylvikt (g/mol):
208.22
Molekylär sammansättning:
C10H12N2O3
InChi Key:
YGPSJZOEDVAXAB-QMMMGPOBSA-N
SMILES:
N[C@@H](CC(=O)C1=CC=CC=C1N)C(O)=O
1
–
5
av
5
Resultat
2
L-Kynurenine, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
L-Kynurenine, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
4
L-Kynurenine, MedChemExpress
MedChemExpress L-Kynurenine is a metabolite of the amino acid L-tryptophan. L-Kynurenine is an aryl hydrocarbon receptor agonist.