Learn More
1,4-Butane sultone, 99+%
Shop All Thermo Scientific Chemicals ProductsDescription
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
CAS Min % | 99.0 |
CAS Max % | 100.0 |
Melting Point | 12.5°C to 15.0°C |
Color | Colorless to Yellow |
Density | 1.3310g/mL |
Boiling Point | 153°C (19.0 HPA) |
Flash Point | 154°C |
Infrared Spectrum | Authentic |
Assay Percent Range | 99% min. (GC) |
Packaging | Glass bottle |
Show More |
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Product Code |
---|---|---|---|
A0470563 | Certificate of Analysis | 17/05/2025 | 107671000,107675000,107670250 |
A0438822 | Certificate of Analysis | 25/08/2023 | 107671000,107675000,107670250 |
A0452566 | Certificate of Analysis | 26/04/2023 | 107671000,107675000,107670250 |
A0393122 | Certificate of Analysis | 15/12/2022 | 107671000,107675000,107670250 |
A0408912 | Certificate of Analysis | 13/12/2022 | 107671000,107675000,107670250 |
Safety and Handling


- 1,4-Butane sultone
- Warning
- Acute toxicity Category 4
- Germ cell mutagenicity Category 2
- H341-Suspected of causing genetic defects.
- H302-Harmful if swallowed.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.