missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ACSM2A Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 22862-1-AP-150UL
This item is not returnable.
View return policy
Description
This gene encodes a mitochondrial acyl-coenzyme A synthetase that is specific for medium chain fatty acids. These enzymes catalyze fatty acid activation, the first step of fatty acid metabolism, through the transfer of acyl-CoA. These enzymes also participate in the glycine conjugation pathway in the detoxification of xenobiotics such as benzoate and ibuprofen. Expression levels of this gene in the kidney may be correlated with kidney function. This gene and its paralog ACSM2B (Gene ID: 348158), both present on chromosome 16, likely arose from a chromosomal duplication event.Specifications
ACSM2A | |
Polyclonal | |
Unconjugated | |
ACSM2A | |
A 923A4.1, ACSM2, ACSM2A, Butyrate CoA ligase 2A, MACS2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
123876 | |
-20°C | |
Liquid |
Western Blot | |
0.23 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q08AH3 | |
ACSM2A | |
ACSM2A Fusion Protein Ag18856 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-4 of
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
ACSM2A Rabbit anti-Human, Polyclonal, Proteintech > 150 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction