Learn More
Advillin Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 20956-1-AP-150UL
This item is not returnable.
View return policy
Description
AVIL is a member of the gelsolin/villin family of actin regulatory proteins. This protein has structural similarity to villin. It binds actin and may play a role in the development of neuronal cells that form ganglia.Specifications
Advillin | |
Polyclonal | |
Unconjugated | |
AVIL | |
ADVIL, advillin, AVIL, DOC6 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
10677, 11567 | |
-20°C | |
Liquid |
Western Blot | |
0.38 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
O75366, O88398 | |
AVIL | |
Advillin Fusion Protein Ag15114 | |
150 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-4 of
Product Content Correction
Product Title
Advillin Rabbit anti-Human, Mouse, Polyclonal, Proteintech > 150 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction