missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ANKRD54 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 25738-1-AP-20UL
This item is not returnable.
View return policy
Description
Ankyrin (ANK) repeats mediate protein-protein interactions in diverse families of proteins. The number of ANK repeats in a protein can range from 2 to over 20. ANK repeats may occur in combinations with other types of domains. The structural repeat unit contains two anti-parallel helices and a beta-hairpin, with repeats stacked in a superhelical arrangement. LIAR, also known as ANKRD54, is a recently identified ANK repeat-containing protein that is predomitly expressed in tissues rich in ciliated cells, such as olfactory sensory neurons and is predicted to be important to cilia. At least three isoforms of LIAR are known to exist.Specifications
ANKRD54 | |
Polyclonal | |
Unconjugated | |
ANKRD54 | |
ANKRD54, ankyrin repeat domain 54 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
129138, 223690 | |
-20°C | |
Liquid |
Western Blot, Immunohistochemistry (Paraffin) | |
0.37 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q6NXT1, Q91WK7 | |
ANKRD54 | |
ANKRD54 Fusion Protein Ag22759 | |
20 μL | |
Primary | |
Mouse, Human | |
Antibody | |
IgG |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-4 of
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
ANKRD54 Rabbit anti-Human, Mouse, Polyclonal, Proteintech > 20 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction