Learn More
Fisherbrand™ Soda Lime Glass Short Stem Funnel
Manufactured from soda lime glass. Fisherbrand™ Soda Lime Glass Short Stem Funnel is less likely to clog when funneling thicker substances.
282.00 SEK - 2310.00 SEK
Specifications
Material | Soda Lime Glass |
---|---|
For Use With (Application) | Ideal for filtering applications in the laboratory |
Product Code | Brand | Diameter (Metric)Top | Diameter (Metric) Stem | Length (Metric) Stem | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric)Top | Diameter (Metric) Stem | Length (Metric) Stem | Price | Quantity & Availability | ||||
10123682
|
Fisherbrand™
00793110 |
100mm | 12mm | 110mm |
626.00 SEK
Pack of 10 |
In Stock
Log in to see stock. |
||||
10082252
|
Fisherbrand™
00793111 |
120mm | 17mm | 120mm |
2310.00 SEK
Pack of 10 |
Estimated Shipment: 15-07-2025
Log in to see stock. |
||||
10710105
|
Fisherbrand™
00793101 |
30mm | 6mm | 35mm |
296.00 SEK
Pack of 10 |
Estimated Shipment: 15-07-2025
Log in to see stock. |
||||
10042222
|
Fisherbrand™
00793102 |
35mm | 7mm | 40mm |
282.00 SEK
Pack of 10 |
In Stock
Log in to see stock. |
||||
10720295
|
Fisherbrand™
00793104 |
45mm | 7mm | 50mm |
367.00 SEK
Pack of 10 |
Estimated Shipment: 18-07-2025
Log in to see stock. |
||||
10730295
|
Fisherbrand™
00793105 |
50mm | 7mm | 50mm |
367.00 SEK
Pack of 10 |
Estimated Shipment: 15-07-2025
Log in to see stock. |
||||
10124352
|
Fisherbrand™
00793106 |
55mm | 8mm | 60mm |
416.00 SEK
Pack of 10 |
In Stock
Log in to see stock. |
||||
10134352
|
Fisherbrand™
00793107 |
60mm | 8mm | 65mm |
432.00 SEK
Pack of 10 |
Estimated Shipment: 22-07-2025
Log in to see stock. |
||||
10082242
|
Fisherbrand™
00793108 |
75mm | 9mm | 80 mm |
468.00 SEK
Pack of 10 |
In Stock
Log in to see stock. |
||||
10767414
|
Fisherbrand™
00793109 |
80mm | 9mm | 85mm |
528.00 SEK
Pack of 10 |
In Stock
Log in to see stock. |
||||
Description
- Manufactured from Soda Lime glass.
- Short stem funnel
- Less likely to clog when funneling thicker substances.
Specifications
Soda Lime Glass | |
Ideal for filtering applications in the laboratory |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
Fisherbrand™ Soda Lime Glass Short Stem Funnel