missing translation for 'onlineSavingsMsg'
Learn More
Learn More
C11orf67 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 20414-1-AP-150UL
This item is not returnable.
View return policy
Description
AAMDC (C11orf67) may play a role in preadipocyte differentiation and adipogenesis.Specifications
C11orf67 | |
Polyclonal | |
Unconjugated | |
AAMDC | |
AAMDC, C11orf67, CK067, PTD015, UPF0366 protein C11orf67 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
28971, 361606, 66273 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q8R0P4, Q9H7C9 | |
AAMDC | |
C11orf67 Fusion Protein Ag14258 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-5 of 12
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
C11orf67 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 150 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction