missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cadmium Solution for ICP, 1g/L, HNO3 Matrix, Fisher Chemical™
Shop All Fisher Chemical Products

Click to view available options
Quantity:
100 mL
250 mL
500 mL
Unit Size:
100mL
250mL
500mL
Specifications
Specifications
Solution Contains | HNO3 |
Concentration | 1 g/L |
Quantity | 100 mL |
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number | Certificate Type | Date | Product Code |
---|---|---|---|
1121384 | Certificate of Analysis | 13/06/2025 | S 010 100 |
1115099 | Certificate of Analysis | 22/05/2025 | S 010 100 |
1090082 | Certificate of Analysis | 05/03/2025 | S 010 100 |
1086803 | Certificate of Analysis | 05/03/2025 | S 010 100 |
1065838 | Certificate of Analysis | 04/03/2025 | S 010 100 |
Safety and Handling

Product Identifier
- Single-element Standard Solution for ICP. Cadmium (Cd) 1000mg/l in HNO3 2%
- Warning
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 3
- H302-Harmful if swallowed.
- H312-Harmful in contact with skin.
- H332-Harmful if inhaled.
- H412-Harmful to aquatic life with long lasting effects.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P264-Wash thoroughly after handling.
- P270-Do not eat, drink or smoke when using this product.
- P271-Use only outdoors or in a well-ventilated area.
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P330-Rinse mouth.
- P362+P364-Take off contaminated clothing and wash it before reuse.
- P501-Dispose of contents/container to .
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Cadmium Solution for ICP, 1g/L, HNO3 Matrix, Fisher Chemical™ >
Spot an opportunity for improvement?Share a Content Correction