Learn More
Thermo Scientific™ Chart Recorders for Ultra-Low Temperature Freezers
Record seven days of temperature performance at temperatures to -150° C.
911.28 SEK - 24065.17 SEK
Product Code | Brand | For Use With (Equipment) | Type | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | For Use With (Equipment) | Type | Price | Quantity & Availability | |||||
11880573
|
Thermo Scientific™
17020 |
HERAfreeze HFU-B, Forma 900, Revco ExF/DxF, TSE/TSD Series | Ink Chart Paper (pack of 50) |
1323.01 SEK
Pack of 50 |
In Stock
Log in to see stock. |
|||||
10298002
|
Thermo Scientific™
201240 |
TSE/TSD Series, Forma 900, HERAfreeze HFU-B, Revco ExF/DxF | Ink Chart Recorder, Field Installed |
22354.37 SEK
Each |
Estimated Shipment: 20-10-2025
Log in to see stock. |
|||||
10715392
|
Thermo Scientific™
201241 |
TSE/TSD Series, Forma 900, HERAfreeze HFU-B, Revco ExF/DxF | Ink Chart Recorder, Factory Installed |
17564.14 SEK
Each |
Estimated Shipment: 18-09-2025
Log in to see stock. |
|||||
10606013
|
Thermo Scientific™
201248 |
HERAfreeze HFU-B, Forma 900, Revco ExF/DxF, TSE/TSD Series | Inkless Chart Recorder, Field Installed |
18248.47 SEK
Each |
In Stock
Log in to see stock. |
|||||
11983927
|
Thermo Scientific™
201249 |
Forma 900, HERAfreeze HFU-B, Revco ExF/DxF, TSE/TSD Series | Inkless Chart Recorder, Factory Installed |
18362.52 SEK
Each |
Estimated Shipment: 18-09-2025
Log in to see stock. |
|||||
10553984
|
Thermo Scientific™
245232 |
Forma 900, HERAfreeze HFU-B, Revco ExF/DxF, TSE/TSD Series | Chart Recorder Pen, Blue | N/A | ||||||
11336351
|
Thermo Scientific™
398223 |
Revco CxF Series (except ULT350 and ULT390) | Ink Chart Recorder, Field Installed |
20301.42 SEK
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11346351
|
Thermo Scientific™
398224 |
Revco CxF 390, 350 | Ink Chart Recorder, Factory Installed |
18362.52 SEK
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11356351
|
Thermo Scientific™
398225 |
Revco CxF 390, 350 | Ink Chart Recorder, Field Installed |
20301.42 SEK
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10601151
|
Thermo Scientific™
6185 |
Thermo Scientific TSU Series, HERAfreeze HFU-T, Revco UxF | Inkless Chart Paper (pack of 50) |
1163.34 SEK
Each |
Estimated Shipment: 17-07-2025
Log in to see stock. |
|||||
16318158
|
Thermo Scientific™
6383-6V |
Forma 88300, HERAfreeze HFU300T, Revco UxF300 | Inkless Chart Recorder, Field Installed |
23038.69 SEK
Each |
Estimated Shipment: 11-09-2025
Log in to see stock. |
|||||
16357468
|
Thermo Scientific™
6383-6W |
Forma 88300, HERAfreeze HFU300T, Revco UxF300 | Inkless Chart Recorder, Field Installed |
23038.69 SEK
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11366381
|
Thermo Scientific™
CR4567 |
TSX Series, Forma FDE, HERAfreeze HDE, Revco RDE and TDE Series (400 box capacity and larger) | Inkless Chart Recorder, Factory Installed |
24065.17 SEK
Each |
Estimated Shipment: 18-09-2025
Log in to see stock. |
|||||
11336391
|
Thermo Scientific™
FCR400R |
TSU400, Revco UxF400 | Inkless Chart Recorder, Field Installed |
23038.69 SEK
Each |
Estimated Shipment: 25-09-2025
Log in to see stock. |
|||||
11346391
|
Thermo Scientific™
FCR567FTH |
Forma 88500, Forma 88600, Forma 88700, TSU500, TSU600 | Inkless Chart Recorder, Field Installed |
23038.69 SEK
Each |
Estimated Shipment: 25-09-2025
Log in to see stock. |
|||||
11356391
|
Thermo Scientific™
FCR567R |
HERAfreeze HFU700T, HERAfreeze HFU600T, Revco UxF500, Revco UxF600, Revco UxF700 | Inkless Chart Recorder, Field Installed |
23038.69 SEK
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11993907
|
Thermo Scientific™
FCR400FT |
Forma 88400, TSU400 | Inkless Chart Recorder, Field Installed |
23038.69 SEK
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15389615
|
Thermo Scientific™
FCR567TSX |
TSX Series | Inkless Chart Recorder, Field Installed |
23038.69 SEK
Each |
Estimated Shipment: 18-09-2025
Log in to see stock. |
|||||
15369615
|
Thermo Scientific™
FCRP567TSX |
TSX Series | Ink Chart Recorder, Field Installed |
23038.69 SEK
Each |
Estimated Shipment: 06-08-2025
Log in to see stock. |
|||||
15788527
|
Thermo Scientific™
FCR400TSXLE |
TSX400 Ultra-low Freezers | Inkless Chart Recorder, Field Installed |
23038.69 SEK
Each |
Estimated Shipment: 19-08-2025
Log in to see stock. |
|||||
15708537
|
Thermo Scientific™
FCR500TSXLE |
TSX500 Ultra-low Freezers | Inkless Chart Recorder, Field Installed |
23038.69 SEK
Each |
Estimated Shipment: 19-08-2025
Log in to see stock. |
|||||
15728537
|
Thermo Scientific™
FCR600TSXLE |
TSX600 Ultra-low Freezers | Inkless Chart Recorder, Field Installed |
23038.69 SEK
Each |
Estimated Shipment: 04-09-2025
Log in to see stock. |
|||||
15738537
|
Thermo Scientific™
FCR700TSXLE |
TSX700 Ultra-low Freezers | Inkless Chart Recorder, Field Installed |
23038.69 SEK
Each |
Estimated Shipment: 18-09-2025
Log in to see stock. |
|||||
15758537
|
Thermo Scientific™
FCRP400TSXLE |
TSX400 Ultra-low Freezers | Ink Chart Recorder, Field Installed |
23038.69 SEK
Each |
Estimated Shipment: 19-08-2025
Log in to see stock. |
|||||
15778537
|
Thermo Scientific™
FCRP500TSXLE |
TSX500 Ultra-low Freezers | Ink Chart Recorder, Field Installed |
23038.69 SEK
Each |
Estimated Shipment: 28-01-2026
Log in to see stock. |
|||||
15798537
|
Thermo Scientific™
FCRP600TSXLE |
TSX600 Ultra-low Freezers | Ink Chart Recorder, Field Installed |
23038.69 SEK
Each |
Estimated Shipment: 19-08-2025
Log in to see stock. |
|||||
15708547
|
Thermo Scientific™
FCRP700TSXLE |
TSX700 Ultra-low Freezers | Ink Chart Recorder, Field Installed |
23038.69 SEK
Each |
Estimated Shipment: 14-08-2025
Log in to see stock. |
|||||
15778527
|
Thermo Scientific™
FCR400LE |
Revco RLE, Forma 89000, TLE Series, HLE Series (400 models only) | Inkless Chart Recorder, Field Installed |
23038.69 SEK
Each |
Estimated Shipment: 18-09-2025
Log in to see stock. |
|||||
15798527
|
Thermo Scientific™
FCR500LE |
Revco RDE, Forma FDE, TDE Series, HDE Series (500 box capacity models only) | Inkless Chart Recorder, Field Installed |
23038.69 SEK
Each |
Estimated Shipment: 07-10-2025
Log in to see stock. |
|||||
15718537
|
Thermo Scientific™
FCR600LE |
Revco RDE, Forma FDE, TDE Series, HDE Series (600 box capacity models only) | Inkless Chart Recorder, Field Installed |
23038.69 SEK
Each |
Estimated Shipment: 03-09-2025
Log in to see stock. |
|||||
15748537
|
Thermo Scientific™
FCRP400LE |
Revco RLE, Forma 89000, TLE Series, HLE Series (400 models only) | Ink Chart Recorder, Field Installed |
23038.69 SEK
Each |
Estimated Shipment: 18-09-2025
Log in to see stock. |
|||||
15768537
|
Thermo Scientific™
FCRP500LE |
Revco RLE, Forma 89000, TLE Series, HLE Series (500 models only) | Ink Chart Recorder, Field Installed |
23038.69 SEK
Each |
In Stock
Log in to see stock. |
|||||
15788537
|
Thermo Scientific™
FCRP600LE |
Revco RLE, Forma 89000, TLE Series, HERAfreeze HLE 600 | Ink Chart Recorder, Field Installed |
23038.69 SEK
Each |
Estimated Shipment: 19-08-2025
Log in to see stock. |
|||||
17888000
|
Thermo Scientific™
332166G01 |
TDE Series Chest Freezers | Inkless Chart Recorder, Factory Installed |
19845.20 SEK
Each |
Estimated Shipment: 04-09-2025
Log in to see stock. |
|||||
17898000
|
Thermo Scientific™
332166G02 |
TDE Series Chest Freezers | Ink Chart Recorder, Factory Installed |
19845.20 SEK
Each |
Estimated Shipment: 04-09-2025
Log in to see stock. |
|||||
17818020
|
Thermo Scientific™
332791G01 |
TDE Series Chest Freezers | Inkless Chart Recorder, Factory Installed |
19845.20 SEK
Each |
Estimated Shipment: 04-09-2025
Log in to see stock. |
|||||
17828020
|
Thermo Scientific™
332791G02 |
TDE Series Chest Freezers | Ink Chart Recorder, Factory Installed |
19845.20 SEK
Each |
Estimated Shipment: 04-09-2025
Log in to see stock. |
|||||
Description
Record seven days of temperature performance at temperatures to -150° C using Thermo Scientific™ Chart Recorders for Ultra-Low Temperature Freezers. Recorders use 6 in. (15cm) chart paper and are available as factory or field installed.
Features:- Seven-day recorder
- Functions to -150°C
- Field or factory installed
Ordering information:
- Field-installed units must be installed by a qualified professional
Specifications
Recorder |