missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CHD9 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 13402-1-AP-20UL
This item is not returnable.
View return policy
Description
The protein encoded by this gene is a 3′-exoribonuclease, with similarity to the RNase D family of 3′-exonucleases. It prefers poly(A) as the substrate, hence, efficiently degrades poly(A) tails of mRNAs. Exonucleolytic degradation of the poly(A) tail is often the first step in the decay of eukaryotic mRNAs. This protein is also involved in silencing of certain maternal mRNAs during oocyte maturation and early embryonic development, as well as in nonsense-mediated decay (NMD) of mRNAs that contain premature stop codons. Alternatively spliced transcript variants encoding different isoforms have been found for this gene.Specifications
CHD9 | |
Polyclonal | |
Unconjugated | |
CHD9 | |
AD013, ATP dependent helicase CHD9, CHD 9, CHD9, CReMM, KIAA0308, KISH2, Kismet homolog 2, PRIC320, x0008 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
109151, 307726, 80205 | |
-20°C | |
Liquid |
Immunofluorescence, Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry | |
0.5 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q3L8U1, Q8BYH8 | |
CHD9 | |
CHD9 Fusion Protein Ag4245 | |
20 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-4 of
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
CHD9 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 20 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction