Learn More
DHRS1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 16275-1-AP-150UL
This item is not returnable.
View return policy
Description
This gene encodes a member of the short-chain dehydrogenases/reductases (SDR) family. The encoded enzyme contains a conserved catalytic domain and likely functions as an oxidoreductase. Multiple alternatively spliced variants, encoding the same protein, have been identified.Specifications
DHRS1 | |
Polyclonal | |
Unconjugated | |
DHRS1 | |
DHRS1, SDR19C1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
115817, 290234, 52585 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.19 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q96LJ7, Q99L04 | |
Dhrs1 | |
DHRS1 Fusion Protein Ag9351 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-4 of
Product Content Correction
Product Title
DHRS1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 150 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction