missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DNAJC7 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 11090-1-AP-20UL
This item is not returnable.
View return policy
Description
DNAJC7 belongs to the evolutionarily conserved DNAJ/HSP40 family of proteins, which regulate molecular chaperone activity by stimulating ATPase activity. DNAJ proteins may have up to 3 distinct domains: a conserved 70-amino acid J domain, usually at the N terminus; a glycine/phenylalanine (G/F)-rich region; and a cysteine-rich domain containing 4 motifs resembling a zinc finger domain (Ohtsuka and Hata, 2000 ).Specifications
DNAJC7 | |
Polyclonal | |
Unconjugated | |
DNAJC7 | |
DANJC7, DJ11, DNAJC7, TPR repeat protein 2, TPR2, TTC2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
56354, 7266 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Flow Cytometry, Western Blot, Immunocytochemistry, Immunofluorescence | |
0.28 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q99615, Q9QYI3 | |
Dnajc7 | |
DNAJC7 Fusion Protein Ag1572 | |
20 μL | |
Primary | |
Mouse, Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
DNAJC7 Rabbit anti-Human, Mouse, Polyclonal, Proteintech > 20 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction