missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EDF1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 12419-1-AP-150UL
This item is not returnable.
View return policy
Description
This gene is known to suppress G-protein and mitogen-activated signal transduction in mammalian cells. The encoded protein shares significant similarity with Arabidopsis FUS6, which is a regulator of light-mediated signal transduction in plant cells. Two alternatively spliced transcript variants encoding different isoforms have been found for this gene.Specifications
EDF1 | |
Polyclonal | |
Unconjugated | |
EDF1 | |
EDF 1, EDF1, MBF1, Multiprotein bridging factor 1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
59022, 8721 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.27 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
O60869, Q9JMG1 | |
Edf1 | |
EDF1 Fusion Protein Ag3063 | |
150 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-4 of
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
EDF1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech > 150 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction