missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ELOVL4 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 55023-1-AP-150UL
This item is not returnable.
View return policy
Description
The antibody is specific to ELOVL4.
This gene encodes a membrane-bound protein which is a member of the ELO family, proteins which participate in the biosynthesis of fatty acids. Consistent with the expression of the encoded protein in photoreceptor cells of the retina, mutations and small deletions in this gene are associated with Stargardt-like macular dystrophy (STGD3) and autosomal domit Stargardt-like macular dystrophy (ADMD), also referred to as autosomal domit atrophic macular degeneration.Specifications
ELOVL4 | |
Polyclonal | |
Unconjugated | |
ELOVL4 | |
ADMD, ELOVL4, STGD2, STGD3 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
315851, 6785, 83603 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Western Blot | |
0.4 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q9EQC4, Q9GZR5 | |
Elovl4 | |
Peptide | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-4 of
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
ELOVL4 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 150 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction