missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GOT1 Mouse anti-Human, Clone: 5A12E10, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 60317-1-IG-20UL
This item is not returnable.
View return policy
Description
GOT1 is a glutamic-oxaloacetic transaminase is a pyridoxal phosphate-dependent enzyme which exists in cytoplasmic and mitochondrial forms, GOT1 and GOT2, respectively. GOT plays a role in amino acid metabolism and the urea and tricarboxylic acid cycles. The two enzymes are homodimeric and show close homology.Specifications
GOT1 | |
Monoclonal | |
0.5 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P17174 | |
GOT1 | |
GOT1 Fusion Protein Ag7012 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |
Flow Cytometry, Western Blot, Immunocytochemistry, Immunohistochemistry (Paraffin), Immunofluorescence | |
5A12E10 | |
Unconjugated | |
GOT1 | |
CAT, GIG18, GOT1, Transaminase A | |
Mouse | |
Protein G | |
RUO | |
2805 | |
-20°C | |
Liquid |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-4 of
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
GOT1 Mouse anti-Human, Clone: 5A12E10, Proteintech > 20 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction