missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KGA Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 20170-1-AP-20UL
This item is not returnable.
View return policy
Description
This antibody is specific to KGA.
Sahai (1983) demonstrated phosphate-activated glutaminase (EC 3.5.1.2) in human platelets. It is the major enzyme yielding glutamate from glutamine. Significance of the enzyme derives from its possible implication in behavior disturbances in which glutamate acts as a neurotransmitter(Prusiner, 1981). High heritability of platelet glutaminase was indicated by studies of Sahai and Vogel (1983) who found an intraclass correlation coefficient of 0.96 for monozygotic twins and 0.53 for dizygotic twins.Specifications
KGA | |
Polyclonal | |
Unconjugated | |
GLS | |
AAD20, GLS, GLS1, GLSK, glutaminase, K glutaminase, KGA, KGA-specific, KIAA0838, L glutamine amidohydrolase | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
14660, 24398, 2744 | |
-20°C | |
Liquid |
Western Blot, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunofluorescence, Immunocytochemistry | |
0.71 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
D3Z7P3, O94925, P13264 | |
Gls | |
KGA-Specific Fusion Protein Ag14002 | |
20 μL | |
Primary | |
Rat, Mouse, Human | |
Antibody | |
IgG |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-4 of
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
KGA Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 20 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction