missing translation for 'onlineSavingsMsg'
Learn More
Learn More
L-Arginine, Hydrochloride, MP Biomedicals™
489.00 SEK - 4170.00 SEK
Chemical Identifiers
MDL Number | MFCD00002635 |
---|---|
Synonym | l-arginine, arginine, l-+-arginine, l-arg, l +-arginine, s-2-amino-5-guanidinopentanoic acid, h-arg-oh, arginina, arginine, l |
PubChem CID | 6322 |
ChEBI | CHEBI:16467 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11373929
|
MP Biomedicals
0210074380 |
100 g |
489.00 SEK
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11383929
|
MP Biomedicals
0210074390 |
500 g |
2400.00 SEK
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11393929
|
MP Biomedicals
0210074391 |
1 kg |
4170.00 SEK
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Chemical Identifiers
MFCD00002635 | |
l-arginine, arginine, l-+-arginine, l-arg, l +-arginine, s-2-amino-5-guanidinopentanoic acid, h-arg-oh, arginina, arginine, l | |
6322 | |
CHEBI:16467 |
Specifications
1119-34-2 | |
0.992 | |
MFCD00002635 | |
ODKSFYDXXFIFQN-UHFFFAOYNA-N | |
2-amino-5-[(diaminomethylidene)amino]pentanoic acid | |
6322 | |
210.662 | |
L-Arginine Hydrochloride |
White | |
C6H14N4O2 | |
l-arginine, arginine, l-+-arginine, l-arg, l +-arginine, s-2-amino-5-guanidinopentanoic acid, h-arg-oh, arginina, arginine, l | |
NC(CCCN=C(N)N)C(O)=O | |
174.20 | |
CHEBI:16467 | |
Crystalline Powder |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
L-Arginine, Hydrochloride, MP Biomedicals™