Learn More
Labconco™ Rotors for CentriVap™ Systems
For use with Labconco CentriVap Systems
4355.00 SEK - 18220.00 SEK
Specifications
Material | Anodized Aluminum |
---|---|
For Use With (Equipment) | CentriVap™ DNA system |
Product Code | Brand | Holds | Weight (English) | Weight (Metric) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Holds | Weight (English) | Weight (Metric) | Price | Quantity & Availability | ||||
10253323
|
Labconco™
7450700 |
132 x 1.5 to 2 mL Microcentrifuge Tube | 2 lb. | 0.9 kg |
4355.00 SEK
Each |
Estimated Shipment: 07-08-2025
Log in to see stock. |
||||
10165462
|
Labconco™
7461900 |
4 Standard or 2 Deep-well Plates | 3 lb. | 1.4 kg |
14360.00 SEK
Each |
Estimated Shipment: 15-09-2025
Log in to see stock. |
||||
10309292
|
Labconco™
7462900 |
72 x 0.5 mL Microcentrifuge Tube or 60 x 1.5 to 2 mL Microcentrifuge Tube | 2 lb. | 0.9 kg |
4515.00 SEK
Each |
Estimated Shipment: 07-08-2025
Log in to see stock. |
||||
10344373
|
Labconco™
7455100 |
40 x 1.5 to 2 mL Microcentrifuge Tubes or 100 x 5 to 6 mL Tubes | 3 lb. | 1.4 kg |
17405.00 SEK
Each |
Estimated Shipment: 15-09-2025
Log in to see stock. |
||||
10155462
|
Labconco™
7455200 |
32 x 40 mL to 50 mL Microcentrifuge Tubes | 3 lb. | 1.4 kg |
16265.00 SEK
Each |
Estimated Shipment: 14-08-2025
Log in to see stock. |
||||
10334373
|
Labconco™
7455000 |
44 x 12 to 17 mL and 24 x 16 to 17 mL Tubes | 4 lb. | 1.8 kg |
18220.00 SEK
Each |
Estimated Shipment: 14-08-2025
Log in to see stock. |
||||
Description
For use with Labconco CentriVap Systems
- Anodized aluminum rotors hold tubes at angle to maximize surface area
- Two hole sizes on rotors 16-317-54 and 16-317-62 to -64 accommodate tubes with different O.D.
- Maximum tube capacity for a given O.D. may vary with overall tube height
- CentriVap DNA Systems and Proteomic come with DNA Rotor (16-317-54); Acid Systems come with Acid-Resistant 12-13 mm Rotor (16-108-450)
To prevent loss of sample during centrifugation, fill tubes only 0.33 to 0.5 full.
Specifications
Anodized Aluminum | |
CentriVap™ DNA system |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
Labconco™ Rotors for CentriVap™ Systems