missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MAGEA4 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 12508-1-AP-150UL
This item is not returnable.
View return policy
Description
This gene is a member of the MAGEA gene family. The members of this family encode proteins with 50 to 80% sequence identity to each other. The promoters and first exons of the MAGEA genes show considerable variability, suggesting that the existence of this gene family enables the same function to be expressed under different transcriptional controls. The MAGEA genes are clustered at chromosomal location Xq28. They have been implicated in some hereditary disorders, such as dyskeratosis congenita. At least four variants encoding the same protein have been found for this gene.Specifications
MAGEA4 | |
Polyclonal | |
Unconjugated | |
MAGEA4 | |
Cancer/testis antigen 1.4, CT1.4, MAGE 4 antigen, MAGE 41, MAGE 41 antigen, MAGE X2, MAGE X2 antigen, MAGE4, MAGE4A, MAGE4B, MAGEA4, melanoma antigen family A, 4, Melanoma associated antigen 4 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
4103 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.17 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P43358 | |
MAGEA4 | |
MAGEA4 Fusion Protein Ag3193 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-4 of
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
MAGEA4 Rabbit anti-Human, Polyclonal, Proteintech > 150 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction