Learn More
MAP1B Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 21633-1-AP-20UL
 View more versions of this product
Product Code. 16851235
  • 1645.54 SEK / 20µL
This item is not returnable. View return policy
Description
Description
The coenzyme NAD and its derivatives are involved in hundreds of metabolic redox reactions and are utilized in protein ADP-ribosylation, histone deacetylation, and in some Ca(2+) signaling pathways. NMNAT.
Specifications
Specifications
MAP1B
Polyclonal
Unconjugated
MAP1B
MAP 1B, MAP1B, MAP5
Rabbit
Antigen Affinity Chromatography
RUO
17755, 4131
-20°C
Liquid
Immunocytochemistry, Western Blot, Immunohistochemistry (Paraffin), Flow Cytometry, Immunofluorescence
0.49 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
P14873, P46821
Map1b
MAP1B Fusion Protein Ag16255
20 μL
Primary
Human, Mouse
Antibody
IgG
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
MAP1B Rabbit anti-Human, Mouse, Polyclonal, Proteintech > 20 μL; Unconjugated