Learn More
Thermo Scientific™ Oxoid™ Novobiocin Antimicrobial Susceptibility discs
Determine the susceptibility of microorganisms using manual AST methods and a full range of high quality, easy-to-use antimicrobial susceptibility testing discs.
Brand:  Thermo Scientific™ CT0038B
215.43 SEK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
 View more versions of this product
Product Code. 10046292
  • 223.00 SEK / Pack of 250
Description
Manually determine the antibiotic susceptibility of microorganisms using Antimicrobial Susceptibility Testing (AST) methods in conjunction with reliable, easy-to-use Thermo Scientific™ Oxoid™ Novobiocin Antimicrobial Susceptibility discs.
Ease of use is provided by shatterproof spring-loaded cartridges, easy-to-read labeling and end-of-cartridge indicators.
disc diffusion not only categorizes resistant, intermediate and sensitive organisms through quantitative results, but also provides a visual indication of the following:
  • Inoculum level
  • Presence of contamination
  • Resistant mutants
  • Beta-lactamase activity
  • Antagonism and synergy between antibiotics
Each cartridge is individually sealed with a desiccant to maintain a moisture level of <2%, ensuring long term stability.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need, and the quality results your laboratory depends on.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Novobiocin
Oxoid Novobiocin Antimicrobial Susceptibility Discs
Antimicrobial Susceptibility Discs
5 Cartridges/Pk.
50 Discs/Cartridge
30μg
NV
Antimicrobial Susceptibility
50 Discs/Cartridge
SDS
Documents Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number6227127 Certificate TypeCertificate of Analysis Date21/02/2025 Catalog NumberCT0038B
Lot Number6109639 Certificate TypeCertificate of Analysis Date10/05/2024 Catalog NumberCT0038B
Lot Number3718401 Certificate TypeCertificate of Analysis Date01/09/2023 Catalog NumberCT0038B
Lot Number3646697 Certificate TypeCertificate of Analysis Date06/04/2023 Catalog NumberCT0038B
Lot Number3466855 Certificate TypeCertificate of Analysis Date19/04/2022 Catalog NumberCT0038B
Promotions
Promotions
Product Content Correction
Product Title
Thermo Scientific™ Oxoid™ Novobiocin Antimicrobial Susceptibility discs > 30μg