missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PLOD3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 11027-1-AP-20UL
This item is not returnable.
View return policy
Description
Forms hydroxylysine residues in -Xaa-Lys-Gly- sequences in collagens. These hydroxylysines serve as sites of attachment for carbohydrate units and are essential for the stability of the intermolecular collagen cross-links.Specifications
PLOD3 | |
Polyclonal | |
Unconjugated | |
PLOD3 | |
LH3, Lysyl hydroxylase 3, PLOD3 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
26433, 288583, 8985 | |
-20°C | |
Liquid |
Immunofluorescence, Western Blot, Immunocytochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
0.25 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
O60568, Q5U367, Q9R0E1 | |
Plod3 | |
PLOD3 Fusion Protein Ag1480 | |
20 μL | |
Primary | |
Mouse, Human, Rat | |
Antibody | |
IgG |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-4 of
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
PLOD3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 20 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction