missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PPAN Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 11006-1-AP-20UL
This item is not returnable.
View return policy
Description
This antibody is raised against the C-terminal of human PPAN and recognize bands around 55 kDa.
The protein encoded by this gene is an evolutionarily conserved protein similar to yeast SSF1 as well as to the gene product of the Drosophila gene peter pan (ppan). SSF1 is known to be involved in the second step of mRNA splicing. Both SSF1 and ppan are essential for cell growth and proliferation. Exogenous expression of this gene was reported to reduce the anchorage-independent growth of some tumor cells. Read-through transcription of this gene with P2RY11/P2Y(11), an adjacent downstream gene that encodes an ATP receptor, has been found. These read-through transcripts are ubiquitously present and up-regulated during granulocyte differentiation.Specifications
PPAN | |
Polyclonal | |
Unconjugated | |
PPAN | |
BXDC3, Peter Pan homolog, peter pan homolog (Drosophila), PPAN, SSF, Ssf 1, SSF1, SSF2, Suppressor of SWI4 1 homolog | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
235036, 298699, 56342 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunofluorescence, Immunocytochemistry | |
0.33 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q91YU8, Q9NQ55 | |
PPAN | |
PPAN Fusion Protein Ag1419 | |
20 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-4 of
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
PPAN Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 20 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction