Learn More
PTGER4 Mouse anti-Human, Unconjugated, Clone: 4A2A12, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 66921-1-IG-20UL
Description
The PTGER4 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human PTGER4. This antibody recognizes human, mouse, rat, pig antigen. The PTGER4 antibody has been validated for the following applications: IF, IHC, WB, ELISA analysis.
Specifications
PTGER4 | |
Monoclonal | |
1000 μg/mL | |
Mouse | |
Primary | |
Human, Mouse, Rat, Pig |
Western Blot, Immunohistochemistry, Immunofluorescence, ELISA | |
4A2A12 | |
Unconjugated | |
20 μL | |
5734 | |
IgG2b |
Product Suggestions
Customers who viewed this item also viewed.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.