missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SCEL Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 11906-1-AP-20UL
This item is not returnable.
View return policy
Description
May function in the assembly or regulation of proteins in the cornified envelope. The LIM domain may be involved in homotypic or heterotypic associations and may function to localize sciellin to the cornified envelope.Specifications
SCEL | |
Polyclonal | |
Unconjugated | |
SCEL | |
SCEL, sciellin | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
8796 | |
-20°C | |
Liquid |
Immunofluorescence, Immunohistochemistry (Paraffin), Immunocytochemistry, Western Blot, Flow Cytometry | |
0.14 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
O95171 | |
SCEL | |
SCEL Fusion Protein Ag2578 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-4 of
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
SCEL Rabbit anti-Human, Polyclonal, Proteintech > 20 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction