Learn More
SHARPIN Mouse anti-Human, Unconjugated, Clone: 3A9E2, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 67177-1-IG-20UL
Description
The SHARPIN antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human SHARPIN. This antibody recognizes human antigen. The SHARPIN antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
SHARPIN | |
Monoclonal | |
1000 μg/mL | |
Mouse | |
Primary | |
Human |
Western Blot, ELISA | |
3A9E2 | |
Unconjugated | |
20 μL | |
81858 | |
IgG2a |
Product Suggestions
Customers who viewed this item also viewed.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.