missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SLC22A15 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 20626-1-AP-20UL
This item is not returnable.
View return policy
Description
Organic ion transporters, such as SLC22A15, transport various medically and physiologically important compounds, including pharmaceuticals, toxins, hormones, neurotransmitters, and cellular metabolites. These transporters are also referred to as amphiphilic solute facilitators (ASFs).Specifications
SLC22A15 | |
Polyclonal | |
Unconjugated | |
SLC22A15 | |
Flipt 1, FLIPT1, PRO34686, SLC22A15 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
242126, 55356 | |
-20°C | |
Liquid |
Western Blot | |
0.41 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q504N2, Q8IZD6 | |
SLC22A15 | |
SLC22A15 Fusion Protein Ag14547 | |
20 μL | |
Primary | |
Mouse, Human | |
Antibody | |
IgG |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-4 of
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
SLC22A15 Rabbit anti-Human, Mouse, Polyclonal, Proteintech > 20 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction