missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SMARCD1 Mouse anti-Human, Clone: 2F2G6, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 66116-1-IG-150UL
This item is not returnable.
View return policy
Description
The protein encoded by this gene is a member of the SWI/SNF family of proteins, whose members display helicase and ATPase activities and which are thought to regulate transcription of certain genes by altering the chromatin structure around those genes. The encoded protein is part of the large ATP-dependent chromatin remodeling complex SNF/SWI and has sequence similarity to the yeast Swp73 protein. Two transcript variants encoding different isoforms have been found for this gene.Specifications
SMARCD1 | |
Monoclonal | |
1.05 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q96GM5 | |
SMARCD1 | |
SMARCD1 Fusion Protein Ag19100 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |
Western Blot | |
2F2G6 | |
Unconjugated | |
SMARCD1 | |
BAF60A, BRG1 associated factor 60A, CRACD1, Rsc6p, SMARCD1, SWI/SNF complex 60 kDa subunit | |
Mouse | |
Protein G | |
RUO | |
6602 | |
-20°C | |
Liquid |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-4 of
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
SMARCD1 Mouse anti-Human, Clone: 2F2G6, Proteintech > 150 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction