missing translation for 'onlineSavingsMsg'
Learn More
Sodium Phosphate Dibasic, MP Biomedicals™ Product Code.: 11410036

Sodium Phosphate Dibasic, MP Biomedicals™
BIOPHARMA

Product Code. p-7083065 Shop All MP Biomedical Products
100 g, 100g
Click to view available options
Quantity:
100 g
500 g
1 kg
Unit Size:
100g
1kg
500g
This item is not returnable. View return policy

Product Code. 11410036

Brand: MP Biomedicals 02199802.1

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Highly hygroscopic and water soluble. Sodium Phosphate Dibasic, MP Biomedicals is a sodium salt of phosphoric acid. This white powder is an important component of running buffer for denaturing gel electrophoresis.

Sodium phosphate dibasic anhydrous

  • Used for preparing phosphate buffer, which in turn is utilized in X-gal staining
  • Induces glomerular calcification as well as other nephrotic syndrome in Sprague-Dawley rats
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7558-79-4
Molecular Formula HNa2O4P
Molecular Weight (g/mol) 141.96
MDL Number MFCD00003496
InChI Key BNIILDVGGAEEIG-UHFFFAOYSA-L
Synonym disodium hydrogen phosphate, disodium phosphate, disodium hydrogenorthophosphate, sodium phosphate dibasic, dibasic sodium phosphate, acetest, soda phosphate, disodium orthophosphate, sodium hydrogen phosphate, phosphoric acid, disodium saltShow MoreShow Less
PubChem CID 24203
ChEBI CHEBI:34683
SMILES [Na+].[Na+].OP([O-])([O-])=O

Specifications

Physical Form Crystalline Powder
Quantity 100 g
Solubility Information Freely soluble in water; insoluble in alcohol
IUPAC Name disodium;hydrogen phosphate
Formula Weight 141.98
Percent Purity 98.0 to 100.5%
Grade USP

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

3 results found
Lot Number Certificate Type Date Product Code
Lot NumberU1122272286 Certificate TypeCertificate of Analysis Date24/03/2023 Product Code02199802.1
Lot NumberU1122272285 Certificate TypeCertificate of Analysis Date24/03/2023 Product Code02199802.1
Lot NumberS6147 Certificate TypeCertificate of Analysis Date17/03/2022 Product Code02199802.1
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Sodium Phosphate Dibasic, MP Biomedicals™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.