missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TFIIE beta Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 11596-1-AP-150UL
This item is not returnable.
View return policy
Description
The general transcription factor IIE (TFIIE) is part of the RNA polymerase II transcription initiation complex, recruiting TFIIH and being essential for promoter clearance by RNA polymerase II. TFIIE is a heterodimer (and sometimes heterotetramer) of alpha and beta subunits. The protein encoded by this gene represents the beta subunit of TFIIE.Specifications
TFIIE beta | |
Polyclonal | |
Unconjugated | |
GTF2E2 | |
FE, GTF2E2, TF2E2, TFIIE B, TFIIE beta | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
2961, 306516, 68153 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Western Blot | |
0.33 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P29084, Q9D902 | |
GTF2E2 | |
TFIIE beta Fusion Protein Ag2136 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-4 of
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
TFIIE beta Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 150 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction